4-Bromo-5-fluoro-1-methyl-1H-pyrazole 95%

Supplier: Ambeed
Warning

A1074595-1G A1074595-250MG
77565-884EA 303 USD
77565-884 77565-886
4-Bromo-5-fluoro-1-methyl-1H-pyrazole 95%
4-Bromo-5-fluoro-1-methyl-1H-pyrazole

Precaución: For research use only.

Formula: C₄H₄BrFN₂
MDL Number: MFCD28520359
CAS Number: 1783510-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR