N-(4-Methanesulfonyl-2-nitrophenyl)-2H-1,3-benzodioxol-5-amine 95%

Supplier: Ambeed
Warning

Synonyms: MSNBA

A1073362-100MG A1073362-250MG
77565-882EA 82.4 USD
77565-882 77619-796
N-(4-Methanesulfonyl-2-nitrophenyl)-2H-1,3-benzodioxol-5-amine 95%
N-(4-Methanesulfonyl-2-nitrophenyl)-2H-1,3-benzodioxol-5-amine

Precaución: For research use only.

Formula: C₁₄H₁₂N₂O₆S
MW: 336.3199 g/mol
MDL Number: MFCD06679104
CAS Number: 852702-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR