Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 98%
Supplier: Ambeed
|
Danger
|
A107313-250MG
A107313-1G
A107313-5G
77565-880EA
46
USD
77565-880
77570-444
77570-446
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III)
Precaución: For research use only.
Formula:
C₃₃H₅₇CoO₆ |
MDL Number:
MFCD00233617 CAS Number: 14877-41-9 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...