3-Nitro-2H-chromen-2-one 95%

Supplier: Ambeed
Warning

A1062978-100MG A1062978-1G A1062978-250MG
77565-860EA 63 USD
77565-860 77783-036 77783-037
3-Nitro-2H-chromen-2-one 95%
3-Nitro-2H-chromen-2-one

Precaución: For research use only.

Formula: C₉H₅NO₄
MDL Number: MFCD00233577
CAS Number: 28448-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR