2-(3-Chlorophenyl)acetaldehyde 95%

Supplier: Ambeed
Warning

A106067-1G A106067-100MG
77565-848EA 176.13 USD
77565-848 77570-354
2-(3-Chlorophenyl)acetaldehyde 95%
2-(3-Chlorophenyl)acetaldehyde

Precaución: For research use only.

Formula: C₈H₇ClO
MW: 154.59 g/mol
MDL Number: MFCD00002892
CAS Number: 41904-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR