(S)-N1-((1H-Benzo[d]imidazol-2-yl)methyl)-N1-(5,6,7,8-tetrahydroquinolin-8-yl)butane-1,4-diamine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
AMD-070 hydrochloride
A105208-250MG
A105208-50MG
77565-836EA
841.51
USD
77565-836
77682-968
(S)-N1-((1H-Benzo[d]imidazol-2-yl)methyl)-N1-(5,6,7,8-tetrahydroquinolin-8-yl)butane-1,4-diamine hydrochloride 98%
(S)-N1-((1H-Benzo[d]imidazol-2-yl)methyl)-N1-(5,6,7,8-tetrahydroquinolin-8-yl)butane-1,4-diamine hydrochloride
Precaución: For research use only.
Formula:
C₂₁H₂₈ClN₅ MW: 385.934 g/mol |
MDL Number:
MFCD23160058 CAS Number: 880549-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...