5-Bromo-3-fluoro-2-methylbenzonitrile 95%

Supplier: Ambeed
Warning

A101408-5G A101408-1G
77565-760EA 615 USD
77565-760 77570-104
5-Bromo-3-fluoro-2-methylbenzonitrile 95%
5-Bromo-3-fluoro-2-methylbenzonitrile

Precaución: For research use only.

Formula: C₈H₅BrFN
MDL Number: MFCD28739741
CAS Number: 1807117-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR