(2R,2'R,3R,3'R)-WingPhos 97%

Supplier: Ambeed
Warning

Synonyms: (2R,2'R,3R,3'R)-4,4'-Di(anthracen-9-yl)-3,3'-di-tert-butyl-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole

A1005931-100MG A1005931-1G A1005931-50MG
77565-750EA 84.46 USD
77565-750 77595-670 77619-578
(2R,2'R,3R,3'R)-WingPhos 97%
(2R,2'R,3R,3'R)-WingPhos

Precaución: For research use only.

Formula: C₅₀H₄₄O₂P₂
MW: 738.831 g/mol
MDL Number: MFCD31630707
CAS Number: 1884680-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR