Biotinyl tyramide 98%

Supplier: Ambeed
Warning

Synonyms: Biotin-Phenol, N-(4-Hydroxyphenethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

A1005416-100MG A1005416-1G A1005416-250MG A1005416-5G
77565-744EA 121 USD
77565-744 77570-044 77619-566 77782-775
Biotinyl tyramide 98%
Biotinyl tyramide

Precaución: For research use only.

Formula: C₁₈H₂₅N₃O₃S
MDL Number: MFCD27953026
CAS Number: 41994-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR