(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-2-cyclopentylacetic acid 97%

Supplier: Ambeed
Warning

A1002329-1G A1002329-5G
77565-736EA 116 USD
77565-736 77570-010
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-2-cyclopentylacetic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-2-cyclopentylacetic acid

Precaución: For research use only.

Formula: C₂₃H₂₅NO₄
MDL Number: MFCD31729554
CAS Number: 187475-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR