Reversible 96-Well and 60-Well Microcentrifuge Tube Racks
Supplier: POLYCARBIN, INC.
R60-01-01 R96-01-01
77565-730CS 55.86 USD
77565-730 77565-732
Reversible 96-Well and 60-Well Microcentrifuge Tube Racks
Racks Tube Racks
These compact, double-sided polypropylene racks are made from 100% circular economy resin.
  • 96- and 60-well options with 5×16 and 5×12 arrays, respectively
  • Holds 0.2 ml tubes on one side, and 1.5 to 2 ml microcentrifuge tubes on the other side
  • Embossed numbers for organizational purposes
  • Autoclavable and freezer safe
These racks accommodate 0.2 ml thin-walled PCR tubes on one side, while the opposite side holds 1.5 to 2 ml microcentrifuge tubes. Racks are available in 60-well and 96-well options.
Polycarbin's reversible PCR tube and microcentrifuge tube racks are perfect for sustainability-minded scientists engaging in benchtop work or freezer storage. These compact, double-sided polypropylene racks are made from 100% post-consumer recycled content, yielding a product with a significantly reduced carbon footprint.