YPD Broth, Powder
Supplier: RPI
Y20090-1000.0
Y20090-250.0
Y20090-500.0
Y20090-5000.0
Y20090-10000.0
Y20090-25000.0
76348-930EA
174.59
USD
76348-930
76348-932
76348-934
76348-936
77510-512
77510-514
YPD Broth, Powder
Media for Microbiology
Dehydrated Media for Microbiology
For maintaining and propagating yeasts in molecular biology applications. The addition of protein and yeast extract promotes better growth. Dissolve 50 grams in 1 liter of distilled water. Sterilize by autoclaving 15 minutes.