Human Recombinant TPX2 (His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
T40-30H-100 T40-30H-1000 T40-30H-20
77564-938EA 618 USD
77564-938 77564-940 77564-942
Human Recombinant TPX2 (His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human TPX2 protein was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. This product can be used for Aurora A activation in vitro.
  • Shipping temperature: Dry ice