Human Recombinant TRIO (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
T16-11G-500 T16-11G-10 T16-11G-50
77564-788EA 2638 USD
77564-788 77564-784 77564-786
Human Recombinant TRIO (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human TRIO (2627-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.