Human Recombinant Tau-441 (P301L, Avi tag) (from E. coli), Biotinylated
Supplier: Sino Biological
T08-56FBN-100 T08-56FBN-1000 T08-56FBN-20
77564-672EA 1328 USD
77564-672 77564-674 77564-676
Human Recombinant Tau-441 (P301L, Avi tag) (from E. coli), Biotinylated
Proteins and Peptides
Recombinant human Tau-441 (P301L) expressed in E. coli cells with an N-terminal Avi tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice