Human Recombinant Tau-441 (244-372) (from E. coli), Biotinylated
Supplier: Sino Biological
T08-55NB-100 T08-55NB-1000 T08-55NB-20
77564-636EA 1148 USD
77564-636 77564-638 77564-640
Human Recombinant Tau-441 (244-372) (from E. coli), Biotinylated
Proteins and Peptides
Recombinant human tag-free Tau-441 (244-372) was expressed in E. coli cells. Then the Tau-441 protein was biotinylated in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice