Human Recombinant Tau-441 (216-391, Avi tag) (from E. coli), Biotinylated
Supplier: Sino Biological
T08-55LNB-100 T08-55LNB-1000 T08-55LNB-20
77564-618EA 1088 USD
77564-618 77564-620 77564-622
Human Recombinant Tau-441 (216-391, Avi tag) (from E. coli), Biotinylated
Proteins and Peptides
Recombinant human Tau-441 (216-391) was expressed in E. coli cells with an N-terminal Avi tag and biotinylated in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice