Human Recombinant Tau-441 (244-372, His tag) (from E. coli)
Supplier: Sino Biological
T08-55H-100 T08-55H-1000 T08-55H-20
77564-582EA 698 USD
77564-582 77564-584 77564-586
Human Recombinant Tau-441 (244-372, His tag) (from E. coli)
Proteins and Peptides
Recombinant human Tau-441 (244-372) was expressed in E. coli cells using an N-terminal His tag.
  • Shipping temperature: Dry ice