Human Recombinant Tau-441 (from E. coli), Biotinylated
Supplier: Sino Biological
T08-54BN-100 T08-54BN-1000 T08-54BN-20
77564-530EA 1148 USD
77564-530 77564-532 77564-534
Human Recombinant Tau-441 (from E. coli), Biotinylated
Proteins and Peptides
Recombinant full length human tag-free Tau-441 was expressed in E. coli cells. Then the Tau-441 protein was biotinylated in vitro after the final chromatography purification.
  • Shipping temperature: Dry ice