Human Recombinant Tau-441, DYRK1A-phosphorylated (from E. coli), Biotinylated
Supplier: Sino Biological
T08-50RNB-100 T08-50RNB-1000 T08-50RNB-20
77564-470EA 1018 USD
77564-470 77564-472 77564-474
Human Recombinant Tau-441, DYRK1A-phosphorylated (from E. coli), Biotinylated
Proteins and Peptides
Recombinant human tag-free Tau-441 was co-expressed with DYRK1A in E. coli cells, phosphorylated by DYRK1A in vivo and in vitro, then the purified protein was biotinylated prior to the final chromatography purification.
  • Shipping temperature: Dry ice