Human Recombinant Tau-441, TTBK1-phosphorylated (from E. coli)
Supplier: Sino Biological
T08-50ON-100 T08-50ON-1000
77564-462EA 1018 USD
77564-462 77564-464
Human Recombinant Tau-441, TTBK1-phosphorylated (from E. coli)
Proteins and Peptides
Recombinant human tag-free Tau-441 was co-expressed with TTBK1 in E. coli cells. The Tau-441 was phosphorylated by TTBK1 in vivo and in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice