Human Recombinant Tau-441, BRSK2-phosphorylated (from E. coli)
Supplier: Sino Biological
T08-50BN-100 T08-50BN-1000 T08-50BN-20
77564-428EA 1018 USD
77564-428 77564-430 77564-432
Human Recombinant Tau-441, BRSK2-phosphorylated (from E. coli)
Proteins and Peptides
Recombinant human tag-free Tau-441 was co-expressed with BRSK2 in E. coli cells. The Tau-441 was phosphorylated by BRSK2 in vivo and in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice