Human Recombinant TIE2 (K855A, GST tag), Unactive (from Baculovirus-Insect Cells)
Supplier: Sino Biological
T04-16BG-100 T04-16BG-1000 T04-16BG-20
77564-332EA 818 USD
77564-332 77564-334 77564-336
Human Recombinant TIE2 (K855A, GST tag), Unactive (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human kinase-dead TIE2 (K855A) (771-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.