Human Recombinant SBK1 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S56-10G-10 S56-10G-50 S56-10G-500
77564-188EA 348 USD
77564-188 77564-190 77564-192
Human Recombinant SBK1 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human SBK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice