Human Recombinant Alpha-synuclein, PLK2-phosphorylated (His tag) (from E. coli)
Supplier: Sino Biological
S22-54H-100 S22-54H-1000 S22-54H-20
77563-714EA 628 USD
77563-714 77563-716 77563-718
Human Recombinant Alpha-synuclein, PLK2-phosphorylated (His tag) (from E. coli)
Proteins and Peptides
Recombinant full length human Alpha-synuclein was expressed in E. coli cells using an N-terminal His tag and was phosphorylated by co-expression with PLK2 in vivo.
  • Shipping temperature: Dry ice