Human Recombinant SNRK (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S16-10G-500 S16-10G-10 S16-10G-50
77563-588EA 2638 USD
77563-588 77563-584 77563-586
Human Recombinant SNRK (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human SNRK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice