Human Recombinant BCR (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
B01-35G-100 B01-35G-1000 B01-35G-20
77554-554EA 818 USD
77554-554 77554-556 77554-558
Human Recombinant BCR (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human BCR (1-695) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.