PCR Plates, 384-Well, Fully Skirted
Supplier: NEST SCIENTIFIC
402901 402902 402903
77508-038CS 440.43 USD
77508-038 77508-040 77508-042
PCR Plates, 384-Well, Fully Skirted
Microplates PCR Plates
Nest dual material PCR plates feature a polycarbonate full-skirted frame design for excellent warp-resistance during thermal cycling and improved mechanical stability for use with robotic handlers.
  • Low binding
  • Polycarbonate frame
  • Frosted polypropylene wells
  • Suitable for use down to -80 °C
  • Plates screened for autofluorescence