Nutrient Agar, Powder

Supplier: RPI

N14100-1000.0 N14100-2000.0 N14100-500.0 N14100-10000.0 N14100-5000.0
76347-206EA 228.63 USD
76347-206 76347-208 76347-210 77510-348 77510-350
Nutrient Agar, Powder
Media for Microbiology Prepared Media for Microbiology

For cultivating a variety of non-fastidious organisms.


  • Ingredients:
  • Beef extract: 3 g/L
  • Pancreatic digest of gelatin: 5 g/L
  • Agar: 15 g/L


Dissolve 23 g in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121 °C for 15 minutes. Mix medium with gentle agitation to avoid bubbles prior to pouring plates. Store at room temperature.


Precaución: Research or further manufacturing use only, not for food or drug use.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR