Luria Broth Miller's Modified
Supplier: RPI
L24080-1000.0 L24080-10000.0 L24080-2000.0 L24080-500.0 L24080-5000.0 L24080-25000.0
76346-604EA 120.88 USD
76346-604 76346-606 76346-608 76346-610 76346-612 77510-300
Luria Broth Miller's Modified
Media for Microbiology Dehydrated Media for Microbiology
For propagation and maintenace of E.Coli.
  • Low salt formula
  • Molecular biology grade
Dissolve 15.5 grams in 1 liter distilled water and adjust the pH to 7.2. Sterilize by autoclaving for 15 minutes.
Precaución: Research or further manufacturing use only, not for food or drug use.