Latex Examination Gloves
Supplier: NEST SCIENTIFIC
901021 901031 901041 901001 901011
77508-948CS 132.6 USD
77508-948 77508-950 77508-952 77508-944 77508-946
Latex Examination Gloves
Gloves
These Natural latex gloves are a type of disposable glove, made 100% out of latex. They are extremely durable and a great barrier against most chemical solutions. Offer protection and excellent performance in a variety of applications and across a broad range of industries.
  • Perfect solution for the majority of work, these gloves offer unrivalled performance compared to similar alternatives
  • Smooth glove surface with textured fingertips ensures strength as well as dexterity
  • Beaded cuff offers drip protection for the forearm and increased strength when donning
  • Significantly more biodegradable and environmentally friendly than nitrile, vinyl and synthetic alternatives
  • Powder free design reduces risk of powder contamination and keeps your hands clean
  • More elastic than nitrile and feature better puncture resistance than vinyl
  • Natural latex offers the benefit of chemical resistance against most acids, bases, chlorine, iodine and formaldehyde
Tensile strength: Minimum 18.0 MPa (Before aging)
Palm thickness: 0.11 mm
Length: Min 230 mm (ASTM)