Glass Bottom Culture Dishes and Plates
Supplier: NEST SCIENTIFIC
801002 801004 801006 801001
77508-928CS 445.32 USD
77508-928 77508-930 77508-932 77508-926
Glass Bottom Culture Dishes and Plates
Culture Dishes Cell Culture Dishes
Glass bottom series are applied in confocal microscope, high resolution microscope, differential interference contrast microscope, polarized light microscope and phase contrast microscope for cell observation.
  • Can be used for live cell observation
  • Special bottom design for easy grip
  • Round cover glass inserts for good appearance
  • Medical adhesive glue to guarantee non-cytotoxicity
  • Sterilized by E-beam, SAL=10⁻⁶
  • Made of high clarity, 100% virgin polystyrene and high clarity glass as bottoms
  • High quality cover glass of standard thickness
  • Withstand temperature
More Product Information