Water, Laboratory Reagent

Supplier: RPI

W20525-10000.0 W20525-4000.0 W20525-55 W20525-5GAL W20525-4X4L
76348-832EA 49.91 USD
76348-832 76348-834 76348-836 76348-838 77510-506
Water, Laboratory Reagent
Water

Laboratory reagent grade. Purified by steam distillation for use as a research laboratory reagent. Not intended or sold for human consumption. 10 Liter size supplied in a Cubitainer with pouring spout.


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: H₂O
Storage Temperature: Room Temperature
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Appearance Clear Liquid
Alkalinity (Total as CaCO3) ≤ 20 ppm
Aluminum (Al) ≤ 0.2 ppm
Antimony (Sb) ≤ 0.006 ppm
Arsenic (As) ≤ 0.01 ppm
Barium (Ba) ≤ 2 ppm
Beryllium (Be) ≤ 0.004 ppm
Boron (B) ≤ 1 ppm
Bromate ≤ 0.010 ppm
Bromide ≤ 0.005 ppm
Cadmium (Cd) ≤ 0.005 ppm
Calcium (Ca) ≤ 2 ppm
Chloride (ASTM D5442) ≤ 50 ppb
Chromium (Cr) ≤ 0.10 ppm
Cobalt (Co) ≤ 0.15 ppm
Conductivity (ASTM D1125) ≤ 5.0 µS/cm
Copper (Cu) ≤ 1 ppm
E.Coli ≤ 1 MPN/100ml
Fluoride ≤ 4 ppm
Foaming Agents ≤ 0.5 mg/l
Hardness (as CaCO3) ≤ 10 mg/l
Heterotrophic Bacteria (F1094) ≤ 100 cfu/10ml
Iron (Fe) ≤ 0.3 ppm
lead (Pb) ≤ 0.015 ppm
Magnesium (Mg) ≤ 0.10 ppm
Manganese (Mn) ≤ 0.05 ppm
Mercury (Hg) ≤ 0.002 ppm
Molybdenum (Mo) ≤ 0.15 ppm
Nickel (Ni) ≤ 0.005 ppm
Nitrate as N ≤ 10 ppm
pH (ASTM D1293) 5.0 - 7.0
Phosphorous (P) ≤ 1 ppm
Potassium (K) ≤ 1 ppm
Resistivity (ASTM D1125) ≥ 0.2 megaohm
Scandium (SC) ≤ 0.15 ppm
Selenium (Se) ≤ 0.05 ppm
Silica (ASTM D4517) No Limit ppb
Silver (Ag) ≤ 0.10 ppm
Sodium (ASTM 6071) ≤ 50 ppb
Standard Plate Count ≤ 500 cfu/ml
Strontium (Sr) ≤ 0.15 ppm
sulfate ≤ 250 ppm
Thallium (Tl) ≤ 0.002 ppm
Tin (sn) ≤ 0.15 ppm
Titanium (Ti) ≤ 0.15 ppm
TOC (ASTM 5997) No Limit ppm
Total Coliform ≤ 1 MPN/100ml
Total Dissolved Solids ≤ 5 mg/L
Total Radium 0.5835 - 1.491 pCi/l
Uranium (U) ≤ 0.030 ppm
Vanadium (V) ≤ 0.15 ppm
Zinc (Zn) ≤ 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR