TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Supplier: RPI
Danger

T26500-10.0 T26500-25.0 T26500-5.0 T26500-50.0
76348-482EA 540.62 USD
76348-482 77510-452 77510-454 77510-456
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₉H₁₆ClO₆P
Storage Temperature: Refrigerator
MDL Number: MFCD00145469
CAS Number: 51805-45-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystals
Function at Reduction (DTNB) ≥ 95 %
Infrared Spectrum Matches reference spectrum
Purity (TLC) ≥ 99 %
Solubility 30% Clear colorless aqueous solution
BSE/TSE Free Conform
Production Method Chemical Synthesis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR