α-Hydroxy-o-tolyl-β-D-glucopyranoside

Supplier: RPI
Warning

Synonyms: 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin

S23200-100.0 S23200-500.0 S23200-1000.0
76348-204EA 431.22 USD
76348-204 76509-146 77510-436
α-Hydroxy-o-tolyl-β-D-glucopyranoside
α-Hydroxy-o-tolyl-β-D-glucopyranoside

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₃H₁₈O₇
MW: 286.28 g/mol
Melting Pt: 196...202 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00006590
CAS Number: 138-52-3
EINECS: 205-331-6

Order Now

Specification Test Results

Appearance White to off white powder
Identification Conforms to Structure
Purity (HPLC) ≥ 98.0 %
Loss on Drying (105°C, 4 hrs) ≤ 0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR