Sodium hydrogen carbonate

Supplier: RPI

S22060-1000.0 S22060-2500.0 S22060-5000.0 S22060-25000.0
76348-088EA 47.33 USD
76348-088 76348-090 76348-092 77510-424
Sodium hydrogen carbonate
Sodium hydrogen carbonate

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: NaHCO₃
Melting Pt: 300 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8

Order Now

Specification Test Results

Appearance White Crystalline Powder
Identification - Sodium No precipitate with K2CO2. Dense precipitate with H2K2O7Sb2
Identification - Bicarbonate Cold Solution unchanged/slight colored
Normal Carbonate Solution does not assume more than a faint pink in color immediately
Insoluble Substances 1:20 Complete and clear
Assay 99.0 - 100.5 %
Loss on Drying ≤ 0.25 %
Limit of Ammonia ≤ 20 ppm
Cadmium ≤ 0.5 ppm
Lead ≤ 0.5 ppm
Arsenic (As) ≤ 1.5 ppm
Mercury (Hg) ≤ 1.0 ppm
Chloride (Cl) ≤ 0.015 %
Limit of Sulfur Compounds ≤ 0.015 %
BSE/TSE Free Conform


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR