Sodium acetate ACS
Supplier: RPI
S22045-500.0
S22045-10000.0
76348-086EA
31.52
USD
76348-086
77510-422
Sodium acetate ACS
Sodium acetate
Precaución: Research or further manufacturing use only, not for food or drug use.
Formula:
H₃CCOONa Melting Pt: >300 °C Storage Temperature: Room Temperature |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 |
Specification Test Results
Appearance | White Powder |
Assay (Titration) | ≥ 99.0 % |
Calcium (Ca) | ≤ 0.005 % |
Chloride Content | ≤ 0.002 % |
Heavy Metals | ≤ 0.001 % |
Infrared Spectrum | Conforms to structure |
Insoluble Matter | ≤ 0.01 % |
Iron (Fe) | ≤ 0.001 % |
Loss on Drying | ≤ 1.0 % |
Magnesium (Mg) | ≤ 0.002 % |
pH (5%, water) @25 °C | 7.0 - 9.2 |
Phosphate | ≤ 0.001 % |
Grade | ACS |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...