Tris acetate

Supplier: RPI
Warning

Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate

T60070-100.0 T60070-1000.0 T60070-500.0 T60070-5000.0
76348-696EA 65.99 USD
76348-696 76348-698 76348-700 77510-480
Tris acetate
Tris acetate

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: NH₂C(CH₂OH)₃·CH₃COOH
MW: 181.1870 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00038951
CAS Number: 6850-28-8
EINECS: 229-939-6

Order Now

Specification Test Results

Appearance White crystalline powder
Heavy Metals ≤0.001%
Moisture ≤0.3%
pH (0.1M Solution in water) 6.0 - 7.0
Purity ≥98.0%
Residue on Ignition ≤0.1%
BSE/TSE Free BSE/TSE Free
Country of Origin Report

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR