Boc-Gly-Gly-Phe-Gly-OH

Supplier: Ambeed

A325761-1G A325761-250MG
77550-902EA 120 USD
77550-902 77616-430
Boc-Gly-Gly-Phe-Gly-OH
Proteins and Peptides

  • InChI Key: PTUJJIPXBJJLLV-AWEZNQCLSA-N


GHS07
Signalword: Warning


Precaución: for Research Use Only

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR