PureClean™ Tyvek® 1421B Autoclavable Stopper Bowl Covers
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
BCTK50 BCTK52 BCTK52-2PLY12 BCTK54 BCTK56 BCTK60 BCTK64 BCTK72 BCTK80 BCTK90 BCTK120 BCTK144 BCTK20 BCTK24 BCTK26 BCTK30 BCTK32 BCTK32-2PLY8 BCTK34 BCTK36 BCTK36-2PLY8 BCTK38 BCTK38-2PLY8 BCTK40 BCTK42 BCTK42-2PLY10 BCTK42-2PLY12 BCTK44 BCTK46 BCTK48
77531-140CS 2545.13 USD
77531-140 77531-158 77531-148 77531-118 77531-142 77531-124 77531-130 77531-128 77531-122 77531-170 77531-166 77531-136 77531-132 77531-146 77531-120 77531-168 77531-162 77531-152 77531-172 77531-134 77531-144 77531-116 77531-174 77531-154 77531-150 77531-160 77531-138 77531-126 77531-164 77531-156
PureClean™ Tyvek® 1421B Autoclavable Stopper Bowl Covers
Covers
PureClean™ autoclave stopper bowl covers are made from 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® on both sides for maximum breathability during sterilization
  • Eliminate the chance of residue deposits on equipment post sterilization
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduce labor costs and simplify the conformance to wrapping SOP's
  • Individually poly bagged
  • Steam sterilization indicator allows autoclaving at 121 to 125 °C
  • Tear- and puncture-resistant, hydrophobic, lint-free
Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilization. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying. After sterilization and after the outer Tyvek® bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed. Remaining in place over the bowl during setup, their use minimizes microbial or particulate contamination risks.
Información para pedidos: Certificates of conformance sent with each shipment.