Lithium chloride
Supplier: RPI
|
Warning
|
L45000-100.0
L45000-1000.0
L45000-500.0
L45000-5000.0
76346-686EA
85.05
USD
76346-686
76346-688
76346-690
77510-306
Lithium chloride
Lithium chloride
Precaución: Research or further manufacturing use only, not for food or drug use.
Formula:
LiCl MW: 42.39 g/mol Boiling Pt: 1382 Melting Pt: 614 Storage Temperature: Room Temperature |
MDL Number:
MFCD00011078 CAS Number: 7447-41-8 EINECS: 231-212-3 Merck Index: 13,05550 |
Specification Test Results
Appearance | White Crystals |
Acid Insolubles | ≤0.01% |
Assay (LiCl) | ≥99.0% |
Calcium (Ca) | ≤0.02% |
Sodium (Na) | ≤0.2% |
Sulfate (SO4) | ≤0.04% |
Water | ≤0.5% |
BSE/TSE Free | BSE/TSE Free |
Country of Origin | Report |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...