D-(-)-Luciferin potassium salt

Supplier: RPI

L37060-0.05 L37060-0.1 L37060-0.25 L37060-1.0 L37060-3.0
76346-660EA 94.9 USD
76346-660 76346-662 76346-664 76346-666 77510-304
D-(-)-Luciferin potassium salt
D(-)-Luciferin potassium salt

The highest purity D-Luciferin Potassium Salt for a wide range of bioluminescent applications. Produces bioluminescence when oxidized by firefly luciferase. Allows highly sensitive bioluminescent measurements. Common research uses reporter gene assays, ATP assays, whole animal studies, DNA sequencing and others. Our product is specially purified and tested extensively to ensure the highest signal and reproducibility with low background and sensitivity. Guaranteed purity is determined by both standard HPLC and Chiral HPLC methods. Protect from light and moisture.


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₁H₇KN₂O₃S₂
Boiling Pt: >300 °C
Melting Pt: >200 °C
Storage Temperature: Freezer
MDL Number: MFCD00044928
CAS Number: 115144-35-9

Order Now

Specification Test Results

Appearance Pale Yellow to Yellow Solid
Chiral Purity ≥97.0%
HPLC (Purity) ≥99%
Identification (FTIR) Conforms to Structure
Impurity Report %
Water ≤9.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR