BD PosiFlush™ Saline Syringes, BD Medical
Supplier: BD
306545 306544 306546
BD306545CS 387.05 USD
BD306545 BD306544 BD306546
BD PosiFlush™ Saline Syringes, BD Medical
Syringes Syringes, Medical
Latex-free with preservative-free saline suitable for use with most patient populations
All syringes feature a unique design to significantly reduce blood reflux and associated line complications.
Regardless of volume, all syringes have the diameter of a standard 10mL syringe, for optimal pressure distribution on the first flush. Solution and fluid path are terminally sterilized to 10-6 Sterility Assurance Level. The stubby syringe profile takes less storage space and lowers sharps waste disposal costs. Clear labeling and a bar code on each syringe helps reduce potential medication errors. Syringes are compatible with all needleless I.V. access systems.