MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt)
Supplier: RPI
Synonyms:
Sodium 2-hydroxy-3-morpholinopropane-1-sulphonate
M81950-1000.0
M81950-2000.0
M81950-250.0
M81950-500.0
77510-334EA
860.56
USD
77510-334
77510-336
77510-338
77510-340
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt)
MOPSO sodium salt (3-(4-Morpholino)-2-hydroxypropanesulphonic acid sodium salt)
Precaución: Research or further manufacturing use only, not for food or drug use.
Formula:
C₇H₁₄NNaO₅S MW: 247.2445 g/mol Storage Temperature: Room Temperature |
MDL Number:
MFCD00070007 CAS Number: 79803-73-9 |
Specification Test Results
Appearance | White crystalline powder |
Solubility (10% in H₂O) | Clear and colorless |
pH | 8.9 - 9.7 |
Absorbance (260 nm) (5% aqueous) | ≤0.04 |
Absorbance (280 nm) (5% aqueous) | ≤0.02 |
Loss on Drying | ≤1.0% |
Assay (by titration) | ≥99.0% |
Residual Solvent (Ethanol Class III) | Complies with ICH Q3C (R5) |
Melamine Free | Melamine Free |
BSE/TSE Free | BSE/TSE Free |
Country of Origin | Report |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...