6-Phosphogluconic acid trisodium salt dihydrate

Supplier: RPI
Warning

Synonyms: D-Glucose-6-phosphate trisodium salt dihydrate, D-Gluconate 6-phosphate trisodium salt dihydrate

P20350-0.1 P20350-0.5 P20350-1.0 P20350-5.0
76347-634EA 65.99 USD
76347-634 76347-636 76347-638 77510-386
6-Phosphogluconic acid trisodium salt dihydrate
6-Phosphogluconic acid trisodium salt dihydrate

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₆H₁₀Na₃O₁₀P·2H₂O
MW: 378.11 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00083681
CAS Number: 57775-17-4
EINECS: 258-543-6

Order Now

Specification Test Results

Appearance White Powder
Solution Test colorless (5% in Water)
Turbidity (0.5% in water) ≤ 2.0 NTU
Water Content (K.F) 8.0 - 12.0 %
Assay 98.0 - 102.0 %
Identity FTIR concordant
1H-NMR Corresponds to assigned structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR