Methylene blue trihydrate

Supplier: RPI
Danger

M25160-100.0 M25160-25.0 M25160-500.0
77510-324EA 105.23 USD
77510-324 77510-326 77510-328
Methylene blue trihydrate
Methylene blue trihydrate

Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₆H₁₈ClN₃S·3H₂O
Melting Pt: 190 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00150008
CAS Number: 7220-79-3

Order Now

Specification Test Results

Appearance Iridescent green-black powder
Ash ≤1.0%
Em (661 nm, Water) ≥69000
Arsenic ≤0.0008%
Copper ≤0.02%
Lead ≤0.001%
Moisture ≤15.0%
Zinc Report %
BSE/TSE Free BSE/TSE Free
Country of Origin Report


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR