PCR Sealing Film
Supplier: NEST SCIENTIFIC
410001 410011 411003
77508-074CS 323.22 USD
77508-074 77508-076 77508-080
PCR Sealing Film
Seals Microplate Seals
NEST PCR sealing film is compatible with NEST 96-well ELISA Plates and 96-well PCR Plates in various applications such as the ELISA/PCR amplification reaction and PCR fluorescence quantification, etc.
  • High transparency PCR sealing film
  • Applicable temperature is −70 to +100 °C
  • Pressure-sensitive film is non-adhesive to skin and gloves, which is convenient for experimental operation and does not affect optical analysis
  • Does not react with the experimental sample, the experimental results are more reliable
  • No autofluorescence
  • Economical and easy to use
  • Compatible with a majority of PCR plates on the market