Toluene-α-sulfonyl fluoride

Supplier: RPI
Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

P20270-1.0 P20270-100.0 P20270-25.0 P20270-250.0 P20270-5.0 P20270-10.0
76347-610EA 20.12 USD
76347-610 76347-612 76347-614 76347-616 76347-618 77510-384
Toluene-α-sulfonyl fluoride
Toluene-α-sulphonyl fluoride

Irreversible serine protease inhibitor of chymotrypsin, kallikrein, papain, subtilisin, thrombin and trypsin. Working conentration is 0.1 to 1.0 mM.


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Melting Pt: 91...94 °C
Storage Temperature: Room Temperature
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2

Order Now

Specification Test Results

Appearance White needle - like crystal
Loss on Drying ≤ 0.5 %
Melting Point 91.0 - 94.0 oC
Purity (GC) ≥ 98.5 %
Solubility Clear, Colorless Solution in Chloroform and Diethyl Ether
TLC Spot One Spot
Non Animal Origin Non Animal Origin
BSE/TSE Free Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR