Nitrofurantoin
Supplier: RPI
|
Danger
|
Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.
Researchers may use nitrofurantoin as a selective agent by incorporating it into culture media to promote the growth of cells or organisms that have been genetically engineered to express resistance to this antibiotic. It may have applications in microbiology and cell culture experiments, particularly in the context of antibiotic selection.
Precaución: Research or further manufacturing use only, not for food or drug use.
Formula:
C₈H₆N₄O₅ Melting Pt: 263...265 °C Storage Temperature: Room Temperature |
MDL Number:
MFCD00003224 CAS Number: 67-20-9 EINECS: 200-646-5 |
Specification Test Results
Appearance | Yellow crystalline powder |
Assay | 98.0 - 102.0 % |
Identification | Pass test |
Sulfated Ash | ≤ 0.1 % |
Related Substances | ≤ 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...