β-Nicotinamide adenine dinucleotide

Supplier: RPI

Synonyms: Coenzyme I, β-NAD, Diphosphopyridine dinucleotide

N30110-1.0 N30110-10.0 N30110-5.0 N30110-25.0
76347-270EA 56.67 USD
76347-270 76347-272 76347-274 77510-358
β-Nicotinamide adenine dinucleotide
β-Nicotinamide adenine dinucleotide

β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.


Precaución: Research or further manufacturing use only, not for food or drug use.

Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4

Order Now

Specification Test Results

Appearance White to off-white powder
Purity ≥ 95 %
NAD Contents Report %
Water Content < 8.0 %
Molar Absorptivity (@260nm - pH 7.5) 17.5 - 18.5 X103L/mole/cm
Ratio @ pH 7.5 A250/A260 0.80 - 0.86
Ratio @ pH 7.5 A280/A260 0.19 - 0.23
Molar Absorptivity when reduced with alcohol dehydrogenase @340nm @ pH10 6.1 - 6.5 X103 L/mole/cm
Ratio when reduced with alcohol dehydrogenase @ pH10 A340/A260 0.42 - 0.44

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR